Learn More
World Precision Instrument Base Weight
SDP
Catalog No. 50822959
Encompass
Sturdy base for the M3301 micromanipulator
This 5-lb weight is for the Tilting Base and provides more stability for your micromanipulator. It is 1"x2.3"x5.1" (25.4 x 60 x 130mm) and has an M6 screw hole for mounting the tilting base.
Catalog No. 50-822-959 Supplier World Precision Instrument Supplier No. 5464
Sturdy base for the M3301 micromanipulator
This 5-lb weight is for the Tilting Base and provides more stability for your micromanipulator. It is 1"x2.3"x5.1" (25.4 x 60 x 130mm) and has an M6 screw hole for mounting the tilting base.