Learn More
Akro Mils Inc TILTVIEW CABINET 4 BINS
Catalog No. NC2209812
Encompass_Preferred
TiltView storage bins tilt out smoothly, and can be removed for cleaning. Crystal clear bin cups allow for quick content identification and close securely for dust protection. These tilting storage bins can be used on a desktop; attached to steel rails, frames or louvered panels; and are wall mountable. TiltView bins are space saving bin units designed to keep parts highly visible and easy to find. Each bin unit comes with card stock labels to identify the bin's contents.Inside dimensions of individual Bin Cups: 4-3/4" D x 4-7/8 W x 7-3/8 H
This item is not returnable. View return policy
Catalog No. NC2209812 Supplier Akro Mils Inc Supplier No. 06704
TiltView storage bins tilt out smoothly, and can be removed for cleaning. Crystal clear bin cups allow for quick content identification and close securely for dust protection. These tilting storage bins can be used on a desktop; attached to steel rails, frames or louvered panels; and are wall mountable. TiltView bins are space saving bin units designed to keep parts highly visible and easy to find. Each bin unit comes with card stock labels to identify the bin's contents.Inside dimensions of individual Bin Cups: 4-3/4" D x 4-7/8 W x 7-3/8 H