Learn More
Medchemexpress LLC Tiludronate disodium | 149845-07-8 | 100.0% | C7H7ClNa2O6P2S | 5 MG
SDP
Catalog No. 5000432580
Encompass_Preferred
Tiludronate disodium is an orally active bisphosphonate that acts as an osteoregulator. It is used for the research of metabolic bone disorders. This compound is a potent inhibitor of the osteoclast vacuolar H(+)-ATPase and possesses antiresorptive and anti-inflammatory properties.
  • Orally active bisphosphonate
  • Acts as an osteoregulator for metabolic bone disorder research
  • Potent inhibitor of osteoclast vacuolar H(+)-ATPase
  • Possesses antiresorptive and anti-inflammatory properties
  • Inhibits proton transport in kidney-derived and osteoclast-derived vesicles
  • Reduces osteoclast capacity to secrete protons and favors detachment from bone matrix
  • Prevents decrease in skeletal mass in osteoporosis models
Catalog No. 50-004-32580 Supplier Medchemexpress LLC Supplier No. HYA0213A5MG
Tiludronate disodium is an orally active bisphosphonate that acts as an osteoregulator. It is used for the research of metabolic bone disorders. This compound is a potent inhibitor of the osteoclast vacuolar H(+)-ATPase and possesses antiresorptive and anti-inflammatory properties.
  • Orally active bisphosphonate
  • Acts as an osteoregulator for metabolic bone disorder research
  • Potent inhibitor of osteoclast vacuolar H(+)-ATPase
  • Possesses antiresorptive and anti-inflammatory properties
  • Inhibits proton transport in kidney-derived and osteoclast-derived vesicles
  • Reduces osteoclast capacity to secrete protons and favors detachment from bone matrix
  • Prevents decrease in skeletal mass in osteoporosis models