Learn More
TARGETMOL CHEMICALS INC TILUDRONATE DISODIUM 5MG
SDP
Catalog No. 502577403
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Tiludronate disodium (Tiludronic acid disodium) is an osteoclast vacuolar H(+)-ATPase inhibitor with antiresorptive and anti-inflammatory properties. Tiludronate disodium can be used in studies about metabolic bone disorders. purity: 100%
Catalog No. 50-257-7403 Supplier TARGETMOL CHEMICALS INC Supplier No. T131595MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Tiludronate disodium (Tiludronic acid disodium) is an osteoclast vacuolar H(+)-ATPase inhibitor with antiresorptive and anti-inflammatory properties. Tiludronate disodium can be used in studies about metabolic bone disorders. purity: 100%