Learn More
Medchemexpress LLC TIM-3/HAVCR2 Human 10ug
SDP
Catalog No. 5000179915
Encompass_Preferred
TIM3 Protein Human (HEK293 Fc) is a type I transmembrane glycoprotein belonging to T Cell Immunoglobulin Mucin (TIM) family and is involved in regulating innate and adaptive immunity
Catalog No. 50-001-79915 Supplier Medchemexpress LLC Supplier No. HYP741510UG
TIM3 Protein Human (HEK293 Fc) is a type I transmembrane glycoprotein belonging to T Cell Immunoglobulin Mucin (TIM) family and is involved in regulating innate and adaptive immunity