Learn More
ACROBiosystems ACROBiosystems Cynomolgus TIM-3 / HAVCR2 Protein, His Tag
Catalog No. 501957209
Encompass_Preferred
Recombinant Protein;100UG;Cynomolgus TIM-3, His Tag (TI3-C52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 201 (Accession # EHH54703.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.6 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus TIM-3, His Tag (Cat. No. TI3-C52H4) at 1 μg/mL (100 μL/well) can bind Anti-TIM-3 MAb with a linear range of 0.8-6 ng/mL (QC tested).
Catalog No. 50-195-7209 Supplier ACROBiosystems Supplier No. TI3C52H4100UG
Recombinant Protein;100UG;Cynomolgus TIM-3, His Tag (TI3-C52H4) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 201 (Accession # EHH54703.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.6 kDa. The protein migrates as 30-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus TIM-3, His Tag (Cat. No. TI3-C52H4) at 1 μg/mL (100 μL/well) can bind Anti-TIM-3 MAb with a linear range of 0.8-6 ng/mL (QC tested).