Learn More
ACROBiosystems Mouse TIM-3 Protein, His Tag, Lyophilized, 100 μg
Catalog No. 501957272
Encompass_Preferred
Hepatitis A virus cellular receptor 2 (HAVCR2), also known as TIM-3, is a Th1-specific cell surface protein that regulates macrophage activation and inhibits T-helper type 1 (Th1)-mediated auto- and alloimmune responses. This mouse TIM-3 protein is expressed from HEK293 cells and contains amino acids Arg 20 to Arg 191. It carries a polyhistidine tag at the C-terminus and is supplied as a lyophilized powder.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 21.0 kDa, migrating as 33-47 kDa under reducing conditions due to glycosylation
  • Endotoxin level of less than 1.0 EU per μg by the LAL method
  • Formulated in PBS, pH 7.4 with trehalose
  • Stable at -20°C to -70°C for 12 months in lyophilized state
Catalog No. 50-195-7272 Supplier ACROBiosystems Supplier No. TI3M52H1100UG
Hepatitis A virus cellular receptor 2 (HAVCR2), also known as TIM-3, is a Th1-specific cell surface protein that regulates macrophage activation and inhibits T-helper type 1 (Th1)-mediated auto- and alloimmune responses. This mouse TIM-3 protein is expressed from HEK293 cells and contains amino acids Arg 20 to Arg 191. It carries a polyhistidine tag at the C-terminus and is supplied as a lyophilized powder.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 21.0 kDa, migrating as 33-47 kDa under reducing conditions due to glycosylation
  • Endotoxin level of less than 1.0 EU per μg by the LAL method
  • Formulated in PBS, pH 7.4 with trehalose
  • Stable at -20°C to -70°C for 12 months in lyophilized state