Learn More
ACROBiosystems ACROBiosystems Human TIM-3 / HAVCR2 Protein, His Tag
Catalog No. 501957023
Encompass_Preferred
Recombinant Protein;1MG;Human TIM-3, His Tag (TM3-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.8 kDa. The protein migrates as 44-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIM-3, His Tag (Cat. No. TM3-H5229) at 0.2 μg/mL (100 μL/well) can bind Anti-TIM3 MAb, Human IgG4 with a linear range of 0.2-6 ng/mL (QC tested).;Anti-Human TIM-3 MAb (human IgG4) captured on CM5 chip via Anti-Human IgG Fc antibodies surface, can bind Human TIM-3, His Tag (Cat. No. TM3-H5229) with an affinity constant of 9.7 nM as determined in a SPR assay (Biacore T200) (Routinely tested).
Catalog No. 50-195-7023 Supplier ACROBiosystems Supplier No. TM3H52291MG
Recombinant Protein;1MG;Human TIM-3, His Tag (TM3-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.8 kDa. The protein migrates as 44-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIM-3, His Tag (Cat. No. TM3-H5229) at 0.2 μg/mL (100 μL/well) can bind Anti-TIM3 MAb, Human IgG4 with a linear range of 0.2-6 ng/mL (QC tested).;Anti-Human TIM-3 MAb (human IgG4) captured on CM5 chip via Anti-Human IgG Fc antibodies surface, can bind Human TIM-3, His Tag (Cat. No. TM3-H5229) with an affinity constant of 9.7 nM as determined in a SPR assay (Biacore T200) (Routinely tested).