Learn More
Enzo Life Sciences TIM-3 monoclonal antibody (1B10) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7020100
Encompass_Preferred
Clone: 1B10. Isotype: IgG1κ. Immunogen: Extracellular domain of human TIM-3. Species reactivity: Human. Applications: Flow Cytometry, ICC, IF, IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide and 50% glycerol.
Catalog No. 50-201-3241