Learn More
AdipoGen anti-Tim-3 (mouse), mAb (TI 142F) (100 µg)
SDP
Supplier:  AdipoGen AG20A0030C100
Encompass_Preferred
Monoclonal Antibody. Recognizes mouse Tim-3. Isotype: Rat IgG2akappa. Clone: TI 142F. Applications: ELISA, FACS. Liquid. 0.2µm-filtered solution in PBS, pH 7.4. Contains no preservatives. Tim-3 is a Th1-specific cell surface protein. It is expressed selectively by differentiated CD4+Th1 and CD8+Tc1 cells, but is absent on CD4+Th2 and CD8+ Tc2 cells. The Tim gene family has an important role in the regulation of autoimmunity and allergies.
Catalog No. 50-205-2812