Learn More
ACROBiosystems Recombinant Protein;Mouse TIM-3 / HAVCR2 Protein, Fc Tag;HEK293;100UG;TI3,HAVCR2,HAVcr-2,KIM3,TIM3,TIMD-3,TIMD3,Tim-3,FLJ14428,KIM-3,KIM3
Catalog No. 501658674
Encompass_Preferred
Recombinant Protein;Mouse TIM-3 / HAVCR2 Protein, Fc Tag;100UG;Mouse TIM-3, Fc Tag (TI3-M5252) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession # AAL65156.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.8 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8674 Supplier ACROBiosystems Supplier No. TI3M5252100UG
Recombinant Protein;Mouse TIM-3 / HAVCR2 Protein, Fc Tag;100UG;Mouse TIM-3, Fc Tag (TI3-M5252) is expressed from human 293 cells (HEK293). It contains AA Arg 20 - Arg 191 (Accession # AAL65156.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.8 kDa. The protein migrates as 60-80 kDa under reducing (R) condition (SDS-PAGE).