Learn More
ACROBiosystems Recombinant Protein;Cynomolgus TIM-3 / HAVCR2 Protein, Fc Tag;HEK293;100UG;TI3,HAVCR2,HAVcr-2,KIM3,TIM3,TIMD-3,TIMD3,Tim-3,FLJ14428,KIM-3,KIM3
Supplier:  ACROBiosystems TI3C525B100UG
Encompass_Preferred
Recombinant Protein;Cynomolgus TIM-3 / HAVCR2 Protein, Fc Tag;100UG;Cynomolgus TIM-3, Fc Tag (TI3-C525b) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 201 (Accession # EHH54703.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 45.8 kDa. The protein migrates as 50-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Cynomolgus TIM-3, Fc Tag (Cat. No. TI3-C525b) at 2μg/mL (100 μL/well) can bind Anti-TIM3 Mab, Human IgG4 with a linear range of 0.5-4 ng/mL (QC tested).
Catalog No. 50-165-8449