Learn More
ACROBiosystems Recombinant Protein;Human TIM-3 / HAVCR2 (22-200) Protein, Fc Tag;HEK293;1MG;TM3,HAVCR2,HAVcr-2,KIM3,TIM3,TIMD-3,TIMD3,Tim-3,FLJ14428,KIM-3,KIM3
Catalog No. 501658345
Encompass_Preferred
Recombinant Protein;Human TIM-3 / HAVCR2 (22-200) Protein, Fc Tag;1MG;Human TIM-3, Fc Tag (TM3-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.5 kDa. The protein migrates as 55-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIM-3 (22-200), Fc Tag (Cat. No. TM3-H5258) at 2 μg/mL (100 µl/well),can bind anti-TIM3 mAb with a linear range of 0.2-3 ng/mL (QC tested).
Catalog No. 50-165-8345 Supplier ACROBiosystems Supplier No. TM3H52581MG
Recombinant Protein;Human TIM-3 / HAVCR2 (22-200) Protein, Fc Tag;1MG;Human TIM-3, Fc Tag (TM3-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 46.5 kDa. The protein migrates as 55-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIM-3 (22-200), Fc Tag (Cat. No. TM3-H5258) at 2 μg/mL (100 µl/well),can bind anti-TIM3 mAb with a linear range of 0.2-3 ng/mL (QC tested).