Learn More
ACROBiosystems Recombinant Protein;Human TIM-3 / HAVCR2 Protein;HEK293;100UG;TM3,HAVCR2,HAVcr-2,KIM3,TIM3,TIMD-3,TIMD3,Tim-3,FLJ14428,KIM-3,KIM3
Supplier:  ACROBiosystems TM3H5229100UG
Encompass_Preferred
Recombinant Protein;Human TIM-3 / HAVCR2 Protein;100UG;Human TIM-3, His Tag (TM3-H5229) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 21.8 kDa. The protein migrates as 44-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIM-3, His Tag (Cat. No. TM3-H5229) at 0.2 μg/mL (100 μL/well) can bind Anti-TIM3 MAb, Human IgG4 with a linear range of 0.2-6 ng/mL (QC tested).
Catalog No. 50-165-8348