Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human TIM-3 / HAVCR2 Protein, His Tag & Avi Tag (Avitag™);HEK293;200UG;TM3,HAVCR2,HAVcr-2,KIM3,TIM3,TIMD-3,TIMD3,Tim-3,FLJ14428,KIM-3,KIM3
Catalog No. 501658323
Encompass_Preferred
Recombinant Protein;Biotinylated Human TIM-3 / HAVCR2 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human TIM-3, His Tag (TM3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 23.1 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-TIM3 MAb, Human IgG4 at 2 μg/mL (100 μL/well) can bind Biotinylated Human TIM-3, His Tag (Cat. No. TM3-H82E7) with a linear range of 0.4-13 ng/mL (QC tested).
Catalog No. 50-165-8323 Supplier ACROBiosystems Supplier No. TM3H82E7200UG
Recombinant Protein;Biotinylated Human TIM-3 / HAVCR2 Protein, His Tag & Avi Tag (Avitag™);200UG;Biotinylated Human TIM-3, His Tag (TM3-H82E7) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 23.1 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-TIM3 MAb, Human IgG4 at 2 μg/mL (100 μL/well) can bind Biotinylated Human TIM-3, His Tag (Cat. No. TM3-H82E7) with a linear range of 0.4-13 ng/mL (QC tested).