Learn More
ACROBiosystems Recombinant Protein;Human TIM-3 / HAVCR2 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;TM3,HAVCR2,HAVcr-2,KIM3,TIM3,TIMD-3,TIMD3,Tim-3,FLJ14428,KIM-3,KIM3
Supplier:  ACROBiosystems TM3H525B100UG
Encompass_Preferred
Recombinant Protein;Human TIM-3 / HAVCR2 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human TIM-3, Mouse IgG2a Fc Tag, low endotoxin (TM3-H525b) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Arg 200 (Accession # Q8TDQ0-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 46.8 kDa. The protein migrates as 50-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human TIM-3, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. TM3-H525b) at 0.2 μg/mL (100 μL/well) can bind Anti-TIM3 MAb, Human IgG4 with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-165-7915