Learn More
Cayman Chemical TIM4 Extracelular DomaInh 20ug
SDP
Catalog No. 502915165
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged TIM-4 extracellular domain expressed in HEK293 cells • Amino acids: 25-315 • MW: 32.7 kDa
Catalog No. 50-291-5165 Supplier Cayman Chemical Supplier No. 3709820 UG
Source: Recombinant human C-terminal His-tagged TIM-4 extracellular domain expressed in HEK293 cells • Amino acids: 25-315 • MW: 32.7 kDa