Learn More
Aviva Systems Biology Corp TIMD4 Antibody - N-terminal region (ARP45554_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45554_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIMD4. It was validated on Western Blot. . Alternate names: FLJ27515, SMUCKLER, TIM4ApplicationWB. Species reactivity: Guinea Pig, Human, Rabbit
Catalog No. 50-285-208