Learn More
Vernier™ Time of Flight Pad
SDPSCI_ED
Catalog No. S15993ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Projectile Launcher
Catalog No. For Use With (Equipment)
S15993ND Projectile Launcher
1 options
Catalog No. S15993ND Supplier Vernier™ Supplier No. TOFVPL
Add to Cart
Add to Cart
Includes
Time of Flight Pad, Cable for connection to Vernier Projectile Launcher, Cable for connection to Vernier interface
Requires
Requires an interface and software, sold separately
Use the Time of Flight Pad to precisely measure how long a projectile has been in motion.
The Time of Flight Pad is electrically like a photogate. When a ball strikes the foam top surface, a circuit is closed.
Warranty and Services
5-year limited warranty
Specifications
For Use With (Application) Educational use only
For Use With (Equipment) Projectile Launcher