Learn More
Research Products International Corp Time Tape, Yellow, 3 Inch Core, 1/2 Inch Wide, 2160 Inch Roll, 6 Rolls per Case
SDP
Supplier:  Research Products International Corp 512604Y
Encompass
Labeling tapes suited for a wide variety of laboratory uses. Easy to write on with pencil, pen, or permanent marker. Strong, durable tapes are resistant to oil, water, and acids. Withstand extreme temperature ranges from -10°F up to +250°F. Can be applied at temperatures as low as +32°F. Features a removable adhesive that peels off glass, plastic or metal.
Catalog No. 50-999-321