Learn More
Aviva Systems Biology Corp TIMELESS Antibody - C-terminal region (ARP38404_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38404_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TIMELESS. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TIMELESS. Alternate names: TIM, TIM1, hTIMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep, Yeast
Catalog No. 50-014-56642