Learn More
Aviva Systems Biology Corp TIMELESS Antibody - N-terminal region (ARP34186_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34186_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TIMELESS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TIMELESS. Alternate names: TIM, TIM1, hTIMApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-55289