Learn More
Andwin Scientific TIMEMED INDCTR TAPE 16/PK
SDP
Catalog No. NC0962058
Encompass_Preferred
Tape is imprinted with the word Autoclaved or Stripes that appear after fifteen minutes of exposure in a steam autoclave set for 250 deg.F. The advanced adhesive assures easy application, strong adhesion and clean removal from the autoclaved items. When autoclaving, shaded dark brown will appear. By coating the double in the Elastomer (Lead compound) ink, and not harmful to health and safety. Without having reached in sterile conditions, diagonal line will appear in the high temperature and a certain amount of time. It is not suitable for use as detection of sterilization conditions.
Catalog No. NC0962058 Supplier Andwin Scientific Supplier No. TSI5342
Tape is imprinted with the word Autoclaved or Stripes that appear after fifteen minutes of exposure in a steam autoclave set for 250 deg.F. The advanced adhesive assures easy application, strong adhesion and clean removal from the autoclaved items. When autoclaving, shaded dark brown will appear. By coating the double in the Elastomer (Lead compound) ink, and not harmful to health and safety. Without having reached in sterile conditions, diagonal line will appear in the high temperature and a certain amount of time. It is not suitable for use as detection of sterilization conditions.