Learn More
TOKU-E TIMENTIN SOLN 10X1ML
Catalog No. NC2402965
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, Ticarcillin Disodium with Clavulanate Potassium 15:1),100 mg/ml in water. Useful in microbiology and plant biology.
Catalog No. NC2402965 Supplier TOKU-E Supplier No. T14710X1ML
Antibiotic, ReadyMade™, sterile-filtered, Ticarcillin Disodium with Clavulanate Potassium 15:1),100 mg/ml in water. Useful in microbiology and plant biology.