Learn More
Bioworld Timentin (Ticarcillin/Clavunate, 15:1), 50 g
Catalog No. NC9734923
Encompass_Preferred
TimentinHighly effective against Gram-negative bacteria such as Agrobacterium strainsA biochemical that inhibits the cross-linking of peptidoglycan during cell wall synthesisIs also a β-Lactamase inhibitorCombination of Ticarcillin disodium and Potassium clavulanateSpecifications:Plant cell culture testedTicarcillin disodium (cas# 4697-14-76)/Potassium clavlanate (cas# 61177-45-5) mixture in a ratio of 15:1HygroscopicFor in vitro lab use only. Not for human use or consumption.Usage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.Appearance: Solid
Catalog No. NC9734923 Supplier Bioworld Supplier No. 420100125
TimentinHighly effective against Gram-negative bacteria such as Agrobacterium strainsA biochemical that inhibits the cross-linking of peptidoglycan during cell wall synthesisIs also a β-Lactamase inhibitorCombination of Ticarcillin disodium and Potassium clavulanateSpecifications:Plant cell culture testedTicarcillin disodium (cas# 4697-14-76)/Potassium clavlanate (cas# 61177-45-5) mixture in a ratio of 15:1HygroscopicFor in vitro lab use only. Not for human use or consumption.Usage: bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY. The product may not be used as a drug, agricultural or pesticidal product , food additive or as a household chemical.Appearance: Solid