Learn More
Bioworld Timentin, 4697-14-76, 61177-45-5, 10g
Catalog No. NC0091860
Encompass_Preferred
Highly effective against Gram-negative bacteria such as Agrobacterium strains. A biochemical that inhibits the cross-linking of peptidoglycan during cell wall synthesis. Is also a β-Lactamase inhibitor. Combination of Ticarcillin disodium and Potassium clavulanate. Specifications: Plant cell culture tested, Ticarcillin disodium (cas# 4697-14-76)/Potassium clavlanate (cas# 61177-45-5) mixture in a ratio of 15:1, Hygroscopic, For in vitro lab use only. Not for human use or consumption. Appearance: Solid. Molecular Weight : 665.64, Molecular Formula : C15H14Na2N2O6S2.C8H8KNO5, Solubility : Soluble in water. Appearance Form : Fine powder. Appearance Color : White. 10g
Catalog No. NC0091860 Supplier Bioworld Supplier No. 420100123
Highly effective against Gram-negative bacteria such as Agrobacterium strains. A biochemical that inhibits the cross-linking of peptidoglycan during cell wall synthesis. Is also a β-Lactamase inhibitor. Combination of Ticarcillin disodium and Potassium clavulanate. Specifications: Plant cell culture tested, Ticarcillin disodium (cas# 4697-14-76)/Potassium clavlanate (cas# 61177-45-5) mixture in a ratio of 15:1, Hygroscopic, For in vitro lab use only. Not for human use or consumption. Appearance: Solid. Molecular Weight : 665.64, Molecular Formula : C15H14Na2N2O6S2.C8H8KNO5, Solubility : Soluble in water. Appearance Form : Fine powder. Appearance Color : White. 10g