Learn More
Alkali Scientific Timentin [Ticarcillin Disodium Salt/Potassium Clavulanate Mixture 15:1 Ratio], 100 G
SDP
Catalog No. 5000660887
Encompass_Preferred
Timentin (100g) combines Ticarcillin Disodium Salt and Potassium Clavulanate in a 15:1 ratio to offer broad-spectrum antibacterial activity. The Ticarcillin component disrupts bacterial cell wall synthesis, while Potassium Clavulanate prevents bacterial resistance by inhibiting beta-lactamase enzymes. This 100g size is ideal for preparing larger volumes of solution for high-throughput testing or clinical treatments. Timentin is highly effective against Gram-negative and Gram-positive organisms, especially those resistant to other antibiotics. It is commonly used in treating complex infections such as sepsis, pneumonia, and intra-abdominal infections in hospital settings.
Catalog No. 50-006-60887 Supplier Alkali Scientific Supplier No. TIM003CP
Timentin (100g) combines Ticarcillin Disodium Salt and Potassium Clavulanate in a 15:1 ratio to offer broad-spectrum antibacterial activity. The Ticarcillin component disrupts bacterial cell wall synthesis, while Potassium Clavulanate prevents bacterial resistance by inhibiting beta-lactamase enzymes. This 100g size is ideal for preparing larger volumes of solution for high-throughput testing or clinical treatments. Timentin is highly effective against Gram-negative and Gram-positive organisms, especially those resistant to other antibiotics. It is commonly used in treating complex infections such as sepsis, pneumonia, and intra-abdominal infections in hospital settings.