Learn More
Alkali Scientific Timentin [Ticarcillin Disodium Salt/Potassium Clavulanate Mixture 15:1 Ratio], 25 G
SDP
Catalog No. 5000660512
Encompass_Preferred
Timentin (25g) is an effective antibiotic combining Ticarcillin Disodium Salt and Potassium Clavulanate in a 15:1 ratio. This formulation increases the effectiveness of Ticarcillin against resistant bacterial strains, especially those producing beta-lactamase. The combination therapy ensures broad-spectrum coverage against a variety of Gram-negative and Gram-positive organisms, including Pseudomonas aeruginosa and Klebsiella pneumoniae. This large-size packaging is ideal for high-volume clinical or laboratory use, especially when large amounts of the drug are required for treating severe infections or in high-throughput testing environments.
Catalog No. 50-006-60512 Supplier Alkali Scientific Supplier No. TIM005CP
Timentin (25g) is an effective antibiotic combining Ticarcillin Disodium Salt and Potassium Clavulanate in a 15:1 ratio. This formulation increases the effectiveness of Ticarcillin against resistant bacterial strains, especially those producing beta-lactamase. The combination therapy ensures broad-spectrum coverage against a variety of Gram-negative and Gram-positive organisms, including Pseudomonas aeruginosa and Klebsiella pneumoniae. This large-size packaging is ideal for high-volume clinical or laboratory use, especially when large amounts of the drug are required for treating severe infections or in high-throughput testing environments.