Learn More
Hach Company Timer, Five-Channel
Supplier:  Hach Company 2630400
Encompass
Four independent channels count up or down for any interval from one second to 100 hours; fifth channel is clock. Large display, one-minute alarm, memory, clock mode. Includes stand, pocket clip, magnet. Dimensions: 7.10 x 6.35 x 1.43 cm (2.875 x 2.50 x 0.56"). Weight: 0.08 kg (2.9 oz). Traceable to NIST.
Catalog No. 50-199-6394