Learn More
Electron Microscopy Sciences Timer, 5-Channel, Certified, Dimensions: 2⅞" x2½" x 9/16". Weight: 2oz
SDP
Catalog No. 501899955
Encompass_Preferred
4 independent channels each of which counts up or down for any interval from 1 second to 100 hours. Its extra large display indicates channels that are in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero. Its memory recalls last timer setting for reuse. Also has a clock mode. Comes complete with a stand, pocket clip, and magnet. Dimensions: 2 7/8" x2 1/2" x 9/16 ". Weight: 2oz.
Catalog No. 50-189-9955 Supplier Electron Microscopy Sciences Supplier No. 7260002
4 independent channels each of which counts up or down for any interval from 1 second to 100 hours. Its extra large display indicates channels that are in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero. Its memory recalls last timer setting for reuse. Also has a clock mode. Comes complete with a stand, pocket clip, and magnet. Dimensions: 2 7/8" x2 1/2" x 9/16 ". Weight: 2oz.