Learn More
Cole-Parmer Traceable Digital Programmable Plug-In Timer Controller with Calibration; 115 VAC
Catalog No. NC0038672 Shop All Cole-Parmer Products
Encompass_Preferred
Plug this controller into any three-prong outlet, and you can turn electrical equipment on or off up to six times a day, seven days a week. The minimum switching interval is one minute. For your convenience, multiple-day switching groups are preprogrammed: Monday-Friday, Monday-Saturday, Saturday-Sunday, and Monday-Sunday. Includes a 24-hour clock and manual program override. Built-in NiCd battery-powered memory retains programs for 10 days in the event of a power failure.
Catalog No. NC0038672 Supplier Cole-Parmer Supplier No. 0864515
Plug this controller into any three-prong outlet, and you can turn electrical equipment on or off up to six times a day, seven days a week. The minimum switching interval is one minute. For your convenience, multiple-day switching groups are preprogrammed: Monday-Friday, Monday-Saturday, Saturday-Sunday, and Monday-Sunday. Includes a 24-hour clock and manual program override. Built-in NiCd battery-powered memory retains programs for 10 days in the event of a power failure.