Learn More
Grainger Sensor Switch Interval Timer Switch, Decorator Style, 12 Hr Max, 1-Pole, 1 ea
Catalog No. 502789775
Encompass_Preferred
The PTS Series electronic auto shut-off timer switches provide a simple, elegant decorator-style lighting control alternative to wall switch occupancy sensors. These units feature six preset countdown timer selections and an on/off push-button, making them ideal for managing line voltage lighting or small motor loads in various applications, including commercial, education, and healthcare environments.
  • Provides six preset countdown timer selections with push-button control
  • Operates on 120/277 V AC or 347 V AC
  • Functions without a neutral wiring connection, ideal for retrofit applications
  • Includes a self-contained relay for switching lighting or small motor loads
  • Features green LED indicators for time status and on/off switch state
  • Equipped with a self-grounding mounting strap
  • Compatible with LED lighting
  • Includes wall plate
Catalog No. 50-278-9775 Supplier Grainger Supplier No. 16A716
The PTS Series electronic auto shut-off timer switches provide a simple, elegant decorator-style lighting control alternative to wall switch occupancy sensors. These units feature six preset countdown timer selections and an on/off push-button, making them ideal for managing line voltage lighting or small motor loads in various applications, including commercial, education, and healthcare environments.
  • Provides six preset countdown timer selections with push-button control
  • Operates on 120/277 V AC or 347 V AC
  • Functions without a neutral wiring connection, ideal for retrofit applications
  • Includes a self-contained relay for switching lighting or small motor loads
  • Features green LED indicators for time status and on/off switch state
  • Equipped with a self-grounding mounting strap
  • Compatible with LED lighting
  • Includes wall plate