Learn More
Aviva Systems Biology Corp Timm13 Antibody - N-terminal region (ARP54905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Timm13. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: D10Ertd378e, Tim9, Timm9ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-975