Learn More
Selleck Chemical LLC Timolol Maleate S4123-1g
SDP
Catalog No. 503111091
Encompass_Preferred
Timolol Maleate (MK-950 (S)-Timolol Maleate) is a non-selective beta-adrenergic receptor antagonist for 1/ 2 with Ki of 1 97 nM/2 0 nM
Catalog No. 50-311-1091 Supplier Selleck Chemical LLC Supplier No. S41231G
Timolol Maleate (MK-950 (S)-Timolol Maleate) is a non-selective beta-adrenergic receptor antagonist for 1/ 2 with Ki of 1 97 nM/2 0 nM