Learn More
Apexbio Technology LLC Timolol Maleate 26921-17-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063359
Encompass_Preferred
Timolol Maleate (CAS 26921-17-5) is the maleate salt form of the (S)-enantiomer of Timolol a potent non-selective -adrenergic receptor antagonist It exhibits high binding affinity toward both 1 and 2 adrenergic receptor subtypes with reported Ki values of approximately 1 97 nM ( 1) and 2 0 nM ( 2) Timolol maleate functions by competitively blocking adrenergic signaling pathways thus modulating sympathetic nervous system activities It is widely utilized in biomedical research to investigate -adrenergic signal transduction mechanisms and associated physiological processes including cardiac function and ophthalmic conditions
Catalog No. 50-000-63359 Supplier Apexbio Technology LLC Supplier No. B13505.1
Timolol Maleate (CAS 26921-17-5) is the maleate salt form of the (S)-enantiomer of Timolol a potent non-selective -adrenergic receptor antagonist It exhibits high binding affinity toward both 1 and 2 adrenergic receptor subtypes with reported Ki values of approximately 1 97 nM ( 1) and 2 0 nM ( 2) Timolol maleate functions by competitively blocking adrenergic signaling pathways thus modulating sympathetic nervous system activities It is widely utilized in biomedical research to investigate -adrenergic signal transduction mechanisms and associated physiological processes including cardiac function and ophthalmic conditions