Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Timolol Maleate 98.0+%, TCI America™
SDP

Catalog No. T2905200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2905200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T2905200MG Supplier TCI America Supplier No. T2905200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Non-selective beta-blocker used for research in pharmacological and pharmaceutical studies.

Chemical Identifiers

CAS 26921-17-5 , 26839-75-8
Molecular Formula C17H28N4O7S
Molecular Weight (g/mol) 432.49
MDL Number MFCD00058356
InChI Key WLRMANUAADYWEA-NWASOUNVSA-N
Synonym tert-butyl-2s-2-hydroxy-3-4-morpholin-4-yl-1,2,5-thiadiazol-3-yl oxy propyl azanium, tert-butyl-2s-2-hydroxy-3-4-4-morpholinyl-1,2,5-thiadiazol-3-yl oxy propyl ammonium, tert-butyl-2s-3-4-morpholin-4-yl-1,2,5-thiadiazol-3-yl oxy-2-oxidanyl-propyl azanium
PubChem CID 6919026
IUPAC Name (2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
SMILES OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1

Specifications

Melting Point 198°C
Color White
Quantity 200 mg
Formula Weight 432.49
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Timolol Maleate
RTECSNumber UA8475000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.