Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Timolol Maleate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607406 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
300 g
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-406 1 g Amber Glass Bottle
18-607-407 5 g Amber Glass Bottle
18-607-408 25 g Amber Glass Bottle
18-607-409 100 g Amber Glass Bottle
18-607-410 300 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-406 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T11341GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

T1134, 26921-17-5, C13H24N4O3S·C4H4O4

Spectrum™ Chemical Timolol Maleate, USP is a non-selective beta-adrenergic receptor antagonist used to treat glaucoma, heart attacks, migraines, and hypertension. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 26921-17-5
Molecular Formula C17H28N4O7S
Molecular Weight (g/mol) 432.49
MDL Number MFCD00058356
InChI Key WLRMANUAADYWEA-NWASOUNVSA-N
IUPAC Name (2S)-1-(tert-butylamino)-3-{[4-(morpholin-4-yl)-1,2,5-thiadiazol-3-yl]oxy}propan-2-ol; (2Z)-but-2-enedioic acid
SMILES OC(=O)\C=C/C(O)=O.CC(C)(C)NC[C@H](O)COC1=NSN=C1N1CCOCC1

Specifications

CAS Max % 1
pH 3.8 to 4.3
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 1 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Timolol Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.