Learn More
Cayman Chemical TImololmaleate 100mg
SDP
Catalog No. 502954260
Encompass_Preferred
A non-selective β-adrenergic receptor antagonist with log Kd values of -8.27, -9.86, and -6.8 for binding to human β1-, β2-, and β3-adrenoceptors, respectively; used to reduce intraocular pressure in research models of ocular hypertension and glaucoma
Catalog No. 50-295-4260 Supplier Cayman Chemical Supplier No. 13974100 MG
A non-selective β-adrenergic receptor antagonist with log Kd values of -8.27, -9.86, and -6.8 for binding to human β1-, β2-, and β3-adrenoceptors, respectively; used to reduce intraocular pressure in research models of ocular hypertension and glaucoma