Learn More
Cayman Chemical Timolol (maleate), Purity: ≥95%, CAS: 26921-17-5, 50 mg
SDP
Catalog No. 502954367
Encompass_Preferred
A non-selective β-adrenergic receptor antagonist with log Kd values of -8.27, -9.86, and -6.8 for binding to human β1-, β2-, and β3-adrenoceptors, respectively; used to reduce intraocular pressure in research models of ocular hypertension and glaucoma
Catalog No. 50-295-4367 Supplier Cayman Chemical Supplier No. 1397450 MG
A non-selective β-adrenergic receptor antagonist with log Kd values of -8.27, -9.86, and -6.8 for binding to human β1-, β2-, and β3-adrenoceptors, respectively; used to reduce intraocular pressure in research models of ocular hypertension and glaucoma