Learn More
Medchemexpress LLC HY-N0810 5mg Medchemexpress, Timosaponin AIII CAS:41059-79-4 Purity:>98%
SDP
Catalog No. 501933726
Encompass_Preferred
Medchemexpress, HY-N0810 5mg Timosaponin AIII CAS:41059-79-4 Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC 50 of 35.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3726 Supplier Medchemexpress LLC Supplier No. HYN08105MG
Medchemexpress, HY-N0810 5mg Timosaponin AIII CAS:41059-79-4 Timosaponin AIII could inhibit acetylcholinesterase (AChE) activity, with an IC 50 of 35.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.