Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Timosaponin B III | 142759-74-8 | 5 MG
SDP

Catalog No. 5000419053
Encompass_Preferred

Timosaponin B III is a major bioactive steroidal saponin isolated from Anemarrhena asphodeloides Bge. It exhibits anti-inflammatory, anti-platelet aggregative, and anti-depressive effects, making it suitable for various research applications.

  • Major bioactive steroidal saponin
  • Isolated from Anemarrhena asphodeloides Bge
  • Exhibits anti-inflammatory effects
  • Exhibits anti-platelet aggregative effects
  • Exhibits anti-depressive effects
  • For research use only
  • Purity: 99.32%
  • Appearance: white to off-white solid

Catalog No. 50-004-19053 Supplier Medchemexpress LLC Supplier No. HYN68065MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Timosaponin B III is a major bioactive steroidal saponin isolated from Anemarrhena asphodeloides Bge. It exhibits anti-inflammatory, anti-platelet aggregative, and anti-depressive effects, making it suitable for various research applications.

  • Major bioactive steroidal saponin
  • Isolated from Anemarrhena asphodeloides Bge
  • Exhibits anti-inflammatory effects
  • Exhibits anti-platelet aggregative effects
  • Exhibits anti-depressive effects
  • For research use only
  • Purity: 99.32%
  • Appearance: white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.