Learn More
Medchemexpress LLC Timosaponin C | 185432-00-2 | 10 MG
SDP
Catalog No. 5000419151
Encompass_Preferred
Timosaponin C is isolated from Rhizoma Anemarrhenae and shows weaker NO inhibition in N9 microglial cells. It is supplied as a high-purity chemical for research applications.
  • High-purity chemical for research applications
  • Isolated from Rhizoma Anemarrhenae
  • Shows weaker NO inhibition in N9 microglial cells
  • Sealed storage at 4°C, away from moisture and light is recommended
  • In solvent, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-19151 Supplier Medchemexpress LLC Supplier No. HYN700610MG
Timosaponin C is isolated from Rhizoma Anemarrhenae and shows weaker NO inhibition in N9 microglial cells. It is supplied as a high-purity chemical for research applications.
  • High-purity chemical for research applications
  • Isolated from Rhizoma Anemarrhenae
  • Shows weaker NO inhibition in N9 microglial cells
  • Sealed storage at 4°C, away from moisture and light is recommended
  • In solvent, store at -80°C for 6 months or -20°C for 1 month