Learn More
Enzo Life Sciences TIMP-1 (human) ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ENZKIT1470001
Encompass_Preferred
The TIMP-1 ELISA kit is a colorimetric immunometric immunoassay kit with results in 2.5 hours. Alternative name: Tissue inhibitor of metalloproteinases-1. Assay Time: 2.5 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Human .
Catalog No. 50-201-3839