Learn More
Enzo Life Sciences TIMP-1 (human neutrophils) (5µg)
SDP
Supplier:  Enzo Life Sciences ALX200426C005
Encompass_Preferred
Specific inhibitor of matrix metalloproteinases (MMPs). It has no activity on mammalian membrane metalloproteinases, procollagen peptidase or on bacterial metalloproteinases such as thermolysin. Specific inhibitor of matrix metalloproteinases (MMPs). It has no activity on mammalian membrane metalloproteinases, procollagen peptidase or on bacterial metalloproteinases such as thermolysin. Alternative name: Tissue inhibitor of metalloproteinase 1. Purity: ≥92% (SDS-PAGE, Western blot). MW: ~28kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.0, containing 200mM sodium chloride, 5mM CaCl2, 1µM ZnCl2, 0.05% BRIJ-35 and 0.05% sodium azide. Source: Isolated from stimulated human neutrophils. The secreted protein consists of 184 amino acids, six disulfide bonds and two glycosylation sites containing N-linked oligosaccharides. Long Term Storage: -80°C.
Catalog No. 50-200-7878