Learn More
Diagnostic Biosystems TIMP-2 TIMP-2
Catalog No. 502407407
Encompass_Preferred
This antibody recognizes a protein of 21kDA, identified as tissue inhibitor of metalloproteinases-2 (TIMP-2). It is closely related to TIMP-1 and shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the protolytic invasiveness of tumor cells and normal placental trophoblast cells.
Catalog No. 50-240-7407 Supplier Diagnostic Biosystems Supplier No. PDM318
This antibody recognizes a protein of 21kDA, identified as tissue inhibitor of metalloproteinases-2 (TIMP-2). It is closely related to TIMP-1 and shows the highest binding affinity to both the latent (pro) and active forms of 72kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMP-2 inhibits the protolytic invasiveness of tumor cells and normal placental trophoblast cells.