Learn More
ACROBiosystems ACROBiosystems Human TIMP-2 Protein, His Tag
Catalog No. 501956703
Encompass_Preferred
Recombinant Protein;1MG;Human TIMP-2, His Tag (TI2-H5223) is expressed from human 293 cells (HEK293). It contains AA Cys 27 - Pro 220 (Accession # NP_003246.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.6 kDa. The protein migrates as 23-26 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-6703 Supplier ACROBiosystems Supplier No. TI2H52231MG
Recombinant Protein;1MG;Human TIMP-2, His Tag (TI2-H5223) is expressed from human 293 cells (HEK293). It contains AA Cys 27 - Pro 220 (Accession # NP_003246.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.6 kDa. The protein migrates as 23-26 kDa under reducing (R) condition (SDS-PAGE).