Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TIMP1 (Human) ELISA Kit

Catalog No. 89100814 Shop All Abnova Corporation Products
Change view
Click to view available options
Conjugate:
Biotin
HRP
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
89-100-814 Biotin
89-025-038 HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-814 Supplier Abnova™ Supplier No. KA3112
Add to Cart
Add to Cart

Sandwich Enzyme Immunoassay for the quantitative measurement of Human TIMP-1

Specifications

Calibration Range 156 to 10000pg/mL
Conjugate Biotin
Cross Reactivity Human
Description TIMP1 (Human) ELISA Kit is intended for the quantitative measurement of human TIMP1.
Detection Limit 5pg/mL
Detection Method Colorimetric
Includes One 96-well plate pre-coated with anti-Human TIMP-1 antibody, Lyophilized TIMP-1 standards (10ng x 2), Sample / Standard diluent buffer (30mL), Biotin conjugated anti-Human TIMP-1 antibody (Concentrated), Dilution: 1:100 (130μL), Antibody diluent buffer (12mL), Avidin-Biotin-Peroxidase Complex (ABC) (Concentrated), Dilution: 1:100 (130μL), ABC diluent buffer (12mL), TMB substrate (10mL), Stop solution (10mL), Wash buffer (25X) (30mL)
Quantity 96 Tests
Regulatory Status For research use only (RUO)
Sample Type Body Fluid, CCS, Plasma, Serum, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store at 4°C for 6 months, or at -20°C for 12 months.
For Use With (Application) Quantification
Product Type TIMP1 (Human) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.