Learn More
ACROBiosystems Recombinant Protein;Human TIMP1 Protein;HEK293;50UG;TI1,TIMP1,TIMP-1,CLGI,EPA,EPO,FLJ90373,HCI,TIMP,TIMP1M,RP1-230G1.3
Catalog No. 501657757
Encompass_Preferred
Recombinant Protein;Human TIMP1 Protein;50UG;Human TIMP-1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Ala 207 (Accession # AAH00866).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24 kDa. The protein migrates as 28-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7757 Supplier ACROBiosystems Supplier No. TI1H522650UG
Recombinant Protein;Human TIMP1 Protein;50UG;Human TIMP-1, His Tag (TI1-H5226) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Ala 207 (Accession # AAH00866).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24 kDa. The protein migrates as 28-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.