Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 7077MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: 3A4. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HeLa cells. Normal Placenta. Breast, Colon, Endometrial, Prostate or Ovarian Carcinoma. Cellular Staining: Cytoplasm and Cell Surface. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: A synthetic peptide from the N-terminus of human TIMP2 protein (aa56-66) (DSGNDIYGNPI). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 21kDa.Chromosome: 17q25. Gene Symbol: TIMP2. Gene ID: 7077. Ensemble: ENSG00000035862. Uniprot: NP_003246.
Catalog No. 50-188-5221