Learn More
Aviva Systems Biology Corp Timp2 Antibody - C-terminal region (ARP42116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Timp2. It was validated on Western Blot. . Alternate names: D11Bwg1104e, Timp-2, Metalloproteinase inhibitor 2, TIMP2, TIMP metallopeptidase inhibitor 2, Tissue inhibitor of metalloproteinases 2, Timp2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-284-977